Skip to Content
Merck
All Photos(1)

Key Documents

AB10320

Sigma-Aldrich

Anti-Cytochrome P450 1B1 Antibody

serum, Chemicon®

Synonym(s):

Cytochrome P450 1B1

Sign Into View Organizational & Contract Pricing

Select a Size

50 μL
€373.00

€373.00


Estimated to ship on14 May 2025


Request a Bulk Order

Select a Size

Change View
50 μL
€373.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

€373.00


Estimated to ship on14 May 2025


Request a Bulk Order

biological source

rabbit

Quality Level

antibody form

serum

antibody product type

primary antibodies

clone

polyclonal

species reactivity

human

manufacturer/tradename

Chemicon®

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... CYP1B1(1545)

General description

Cytochrome P450 1B1 (CYP1B1) is active in the metabolism of estrogens to reactive catechols and of different procarcinogens. The cytochrome P450 proteins are monooxygenases which catalyze many reactions involved in drug metabolism and the synthesis of cholesterol, steroids and other lipids.

Immunogen

Recombinant human 1B1

Application

Suggested dilutions:
Western Blot: 1:5000
Anti-1B1 Antibody detects level of 1B1 & has been published & validated for use in WB.
Research Category
Metabolism
Research Sub Category
Enzymes & Biochemistry

Quality

Routinely evaluated by immunoblot.

Target description

61kDa

Physical form

Raw rabbit serum with 0.05% sodium azide.

Storage and Stability

Maintain at -20°C for up to 12 months from date of receipt.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service