Skip to Content
Merck
All Photos(1)

Key Documents

9510-OP

Millipore

Urea

OmniPur®

Synonym(s):

Urea, Carbamide, Carbonyldiamide

Sign Into View Organizational & Contract Pricing

Select a Size

500 G
Inquire
5 KG
Inquire


Estimated to ship on24 May 2025


Request a Bulk Order

Select a Size

Change View
500 G
Inquire
5 KG
Inquire

About This Item

Linear Formula:
NH2CONH2
CAS Number:
Molecular Weight:
60.06
Beilstein:
635724
MDL number:
UNSPSC Code:
12352111
NACRES:
NA.28


Estimated to ship on24 May 2025


Request a Bulk Order

Quality Level

form

powder

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

color

white

mp

132-135 °C (lit.)

density

1.335 g/mL at 25 °C (lit.)

shipped in

ambient

SMILES string

NC(N)=O

InChI

1S/CH4N2O/c2-1(3)4/h(H4,2,3,4)

InChI key

XSQUKJJJFZCRTK-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description



Ideal for protein unfolding/denaturation. Molecular biology grade.

Specifications:
DNase: None detected
Protease: None detected
RNase: None detected
Assay: ≥99.5%
A260 (0.1 M in water): ≤0.01
A280 (0.1 M in water): ≤0.01

Heavy metals (as Pb): ≤0.001





Intended for laboratory and manufacturing use only. Not for drug, food, or household use.

Application

Used for the denaturation of proteins and as a mild solubilization agent for insoluble or denatured proteins. Useful for renaturing proteins from samples already denatured with 6 M guanidine chloride such as inclusion bodies. May be used with guanidine hydrochloride and dithiothreitrol (DTT) in the refolding of denatured proteins into their native or active form.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany
OMNIPUR is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service