Skip to Content
Merck
All Photos(1)

Key Documents

9480-OP

Millipore

Polysorbate 20

OmniPur®

Synonym(s):

TWEEN® 20, Polyoxyethylene Sorbitan Monolaurate, TWEEN 20, Polyethylene glycol sorbitan monolaurate, Polyoxyethylenesorbitan monolaurate

Sign Into View Organizational & Contract Pricing

Select a Size

1 L
Inquire
4 L
Inquire


Estimated to ship on30 May 2025


Request a Bulk Order

Select a Size

Change View
1 L
Inquire
4 L
Inquire

About This Item

CAS Number:
MDL number:
UNSPSC Code:
12161902
NACRES:
NA.28


Estimated to ship on30 May 2025


Request a Bulk Order

Product Name

OmniPur®Polyoxyethylene (20) Sorbitan Monolaurate,

form

viscous liquid

Quality Level

mol wt

~1228

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

color

lightdark yellow

refractive index

n20/D 1.468 (lit.)

density

1.095 g/mL at 25 °C (lit.)

shipped in

ambient

SMILES string

CCCCCCCCCCCC(=O)OCCOCC(C1C(C(CO1)OCCO)OCCO)OCCO

InChI

1S/C26H50O10/c1-2-3-4-5-6-7-8-9-10-11-24(30)34-19-18-31-20-22(32-15-12-27)26-25(35-17-14-29)23(21-36-26)33-16-13-28/h22-23,25-29H,2-21H2,1H3

InChI key

HMFKFHLTUCJZJO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Non-ionic detergent suited for solubilization of membrane proteins, enzymology, analytical, and diagnostic work, as well as chromatography and electrophoresis. Molecular Biology Grade.

DNAse: none detected
Heavy metals (as Pb): ≤0.001%
Hydroxyl number: 96-108
Residue after ignition: ≤0.25%
Moisture (H2O): ≤3.0%
Protease: none detected
RNAse: none detected





Intended for laboratory and manufacturing use only. Not for drug, food, or household use.

Application

Non-ionic detergent

Warning

Toxicity: Irritant (B)

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany
OMNIPUR is a registered trademark of Merck KGaA, Darmstadt, Germany
TWEEN is a registered trademark of Croda International PLC

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

527.0 °F - Pensky-Martens closed cup

Flash Point(C)

275 °C - Pensky-Martens closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service