Skip to Content
Merck
All Photos(1)

Documents

9060-HK

Sigma-Aldrich

Mouse/Human Adiponectin

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116012
eCl@ss:
32160405
NACRES:
NA.84

conjugate

125Iodine

Quality Level

label

125Iodine labeled

NCBI accession no.

UniProt accession no.

Gene Information

human ... ADIPOQ(9370)
mouse ... ADIPOQ(11450)

General description

Radioactivity: <3 μCi

Application

Research Category
All
appropriate label hydrating buffer if kit replacement or assay buffer of choice

Storage and Stability

2–8 °C

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Chronic 3

Storage Class Code

7 - Radioactive Substances

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service