Skip to Content
Merck
All Photos(1)

Key Documents

8820-OP

Millipore

TBE Buffer

OmniPur® Grade, 10X

Synonym(s):

Tris-Borate-EDTA Buffer

Sign Into View Organizational & Contract Pricing

Select a Size

4 L
Inquire


Estimated to ship on28 May 2025


Request a Bulk Order

Select a Size

Change View
4 L
Inquire

About This Item

UNSPSC Code:
41105319
NACRES:
NA.28


Estimated to ship on28 May 2025


Request a Bulk Order

Quality Level

form

liquid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

color

clear colorless

shipped in

ambient

storage temp.

room temp

General description

Suitable for a wide range of molecular biology applications. Each liter of 10X TBE buffer yields 10 liters of 1X TBE buffer. Final concentration of 1X TBE buffer: 0.089 M Tris, 0.089 M Borate, 0.002 M EDTA. Molecular biology grade.

Specifications
DNAse: none detected
RNAse: none detected
Protease: none detected






Intended for laboratory and manufacturing use only. Not for drug, food, or household use.

Application


  • Horseradish peroxidase-catalyzed polyacrylamide gels: monitoring their polymerization with BSA-stabilized gold nanoclusters and their functional validation in electrophoresis.: This study demonstrates the use of TBE buffer in the polymerization of polyacrylamide gels for electrophoresis, enhancing the resolution and stability of protein and nucleic acid separation, crucial for biochemical analysis (Wang D et al., 2024).

  • A method for high-concentration agarose gel preparation and its application in high-resolution separation of low-molecular-weight nucleic acids and proteins.: Explores the effectiveness of TBE buffer in preparing high-concentration agarose gels, significantly improving the separation efficiency of small biomolecules, which is vital for molecular biology research (Tong Z et al., 2023).

  • Nanocellulose for gel electrophoresis.: Details the role of TBE buffer in developing nanocellulose-based gels for electrophoresis, highlighting its potential to replace traditional matrices with more environmentally friendly and efficient alternatives (Garnier G et al., 2019).

Warning

Toxicity: Irritant (B)

Physical form

Supplied as a 10X solution in water.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany
OMNIPUR is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Repr. 1B

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 2


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH SVHC Candidate List

CAS No.

EU REACH Annex XVII (Restriction List)

CAS No.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service