Skip to Content
Merck
All Photos(1)

Key Documents

8.57013

Sigma-Aldrich

Oxidizing Reagent B for DNA Synthesis

1-Methylimidazole : Tetrahydrofuran 18/82 (w/w), Novabiochem®

Synonym(s):

0.02M Iodine in THF : Pyridine : Water 69/22/11 (w/w/w), Novabiochem®

Sign Into View Organizational & Contract Pricing

Select a Size

4 L
€519.00

€519.00


Estimated to ship on18 May 2025


Request a Bulk Order

Select a Size

Change View
4 L
€519.00

About This Item

UNSPSC Code:
12352202
NACRES:
NA.22

€519.00


Estimated to ship on18 May 2025


Request a Bulk Order

Quality Level

product line

Novabiochem®

form

liquid

autoignition temp.

215 °C (THF)

reaction suitability

reaction type: Oxidations
reagent type: oxidant

manufacturer/tradename

Novabiochem®

transition temp

flash point -21.5 °C (THF)

storage temp.

2-30°C

General description

Pre-mixed ready-to-use Capping Reagent
  • 857013.0450: Compatible with MerMade DNA Synthesisers. Attaches directly to Instrument (28/405 neck finish)
  • 857013.4000: Compatible with MerMade DNA Synthesisers. Stand alone from Instrument (38mm Neck Finish)

Legal Information

Novabiochem is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Oral - Carc. 2 - Eye Irrit. 2 - Flam. Liq. 2 - Skin Irrit. 2

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

-6.7 °F - (THF)

Flash Point(C)

-21.5 °C - (THF)


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service