Skip to Content
Merck
All Photos(1)

Key Documents

8.56186

Sigma-Aldrich

Fmoc-Lys(ivDde)-Wang resin (100-200 mesh)

Novabiochem®

Synonym(s):

Fmoc-Protected Lysine Resin

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
€136.00
5 G
€522.00

€136.00


Estimated to ship on18 May 2025


Request a Bulk Order

Select a Size

Change View
1 G
€136.00
5 G
€522.00

About This Item

UNSPSC Code:
12352209
NACRES:
NA.22

€136.00


Estimated to ship on18 May 2025


Request a Bulk Order

Quality Level

form

beads

manufacturer/tradename

Novabiochem®

functional group

Fmoc

storage temp.

2-8°C

General description

Useful tool for the synthesis of side-chain modified, cyclic and branched peptides by Fmoc SPPS. Following chain extension, the side-chain ivDde group is removed with 2% hydrazine in DMF, enabling selective modification of the ε-lysine amino group.[1]

Associated Protocols and Technical Articles
Cleavage and Deprotection Protocols for Fmoc SPPS
SelectingOrthogonal Building Blocks

Literature references:
[1] R. Chhabra, et al. (1998) Tetrahedron Lett., 39, 1603.

Application

Fmoc-Lys(ivDde)-Wang resin (100-200 mesh) has been used as a solid support to prepare a peptide bearing a hexacarbonyldiiron cluster[2], neoglycopeptides[3] and lipopeptides[4] as putative antifungal and cancer vaccines, respectively.

Analysis Note

Color (visual): white to yellow to beige
Appearance of substance (visual): beads
Loading (Photometric determination of the Fmoc-chromophore liberated upon treatment with DBU/DMF): 0.40 - 1.00 mmol/g
Swelling Volume (in DMF): lot specific result
Identity (of the substitution): passes test
The polymer matrix is copoly (styrene -1 % DVB), 100- 200 mesh.

Legal Information

Novabiochem is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

An appraisal of new variants of Dde amine protecting group for solid phase peptide synthesis
R. Chhabra, et al.
Tetrahedron Letters, 39, 1603-1603 (1998)
Towards a fully synthetic MUC1-based anticancer vaccine: Efficient conjugation of glycopeptides with mono-, di-, and tetravalent lipopeptides using click chemistry
Cai H, et al.
Chemistry?A European Journal , 17, 6396-6406 (2011)
Artificial [FeFe]-Hydrogenase: On Resin Modification of an Amino Acid to Anchor a Hexacarbonyldiiron Cluster in a Peptide Framework
Roy S, et al.
European Journal of Inorganic Chemistry, 1050-1055 (2011)
Jonathan Cartmell et al.
Carbohydrate research, 403, 123-134 (2014-08-16)
Selective strategies for the construction of novel three component glycoconjugate vaccines presenting Candida albicans cell wall glycan (β-1,2 mannoside) and polypeptide fragments on a tetanus toxoid carrier are described. The first of two conjugation strategies employed peptides bearing an N-terminal

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service