Select a Size
| Size/SKU | Availability | Price |
|---|---|---|
1 g | Check Cart for Availability | € 111,00 |
5 g | Check Cart for Availability | € 437,00 |
About This Item
Quality Segment
product line
Novabiochem®
form
beads
reaction suitability
reaction type: Fmoc solid-phase peptide synthesis
manufacturer/tradename
Novabiochem®
application(s)
peptide synthesis
functional group
Fmoc
storage temp.
2-30°C
General description
Associated Protocols and Technical Articles
Cleavage and Deprotection Protocols for Fmoc SPPS
Analysis Note
Appearance of substance (visual): beads
Loading (photometric determination of the Fmoc-chromophore liberated upon treatment with piperidine/DMF.): 0.40 - 1.00 mmol/g
Swelling Volume (in DMF): lot specific result
Identity (of the substitution): passes test
The polymer matrix is copoly (styrene-1 % DVB), 200 - 400 mesh.
Other Notes
Legal Information
1 of 1
This Item | |||
|---|---|---|---|
| reaction suitability reaction type: Fmoc solid-phase peptide synthesis | reaction suitability reaction type: Fmoc solid-phase peptide synthesis | reaction suitability reaction type: Fmoc solid-phase peptide synthesis | reaction suitability reaction type: Fmoc solid-phase peptide synthesis |
| functional group Fmoc | functional group Fmoc | functional group Fmoc | functional group Boc, Fmoc |
| product line Novabiochem® | product line Novabiochem® | product line Novabiochem® | product line Novabiochem® |
| Quality Level 200 | Quality Level 200 | Quality Level 200 | Quality Level 200 |
| application(s) peptide synthesis | application(s) peptide synthesis | application(s) peptide synthesis | application(s) peptide synthesis |
| form beads | form beads | form beads | form beads |
Storage Class
11 - Combustible Solids
wgk
WGK 1
flash_point_f
Not applicable
flash_point_c
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.



