Skip to Content
Merck
All Photos(1)

Key Documents

8.55131

Sigma-Aldrich

Dawson Dbz AM resin (100-200 mesh)

3-(Fmoc-amino)-4-aminobenzoyl AM resin (100-200 mesh) Novabiochem®

Synonym(s):

3-(Fmoc-amino)-4-aminobenzoyl Rink Amide AM resin (100 - 200 mesh), Dawson Nbz AM resin (100-200 mesh)

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
€91.20
5 G
€364.00
25 G
€1,470.00

€91.20


Estimated to ship on25 May 2025


Request a Bulk Order

Select a Size

Change View
1 G
€91.20
5 G
€364.00
25 G
€1,470.00

About This Item

UNSPSC Code:
13111023
NACRES:
NA.22

€91.20


Estimated to ship on25 May 2025


Request a Bulk Order

Quality Level

product line

Novabiochem®

form

beads

reaction suitability

reaction type: Fmoc solid-phase peptide synthesis

manufacturer/tradename

Novabiochem®

application(s)

peptide synthesis

functional group

amine

storage temp.

2-8°C

General description

A novel safety-catch resin for preparation peptide thioesters by Fmoc SPPS. Linker is activated to cleavage by thiols by treatment with CDI.

Associated Protocols and Technical Articles
Protocols for Loading of Peptide Synthesis Resins

Analysis Note

Color (visual): light yellow to yellow to brown
Appearance of substance (visual): beads
Loading (photometric determination of the Fmoc-chromophore liberated upon treatment with piperidine/DMF.): 0.40 - 0.80 mmol/g
Swelling Volume (in DMF): lot specific result
Swelling Volume (in CH₂Cl₂): lot specific result
Total swelling volume acc. Houben Weyl (in CH2Cl2): ≥ 5.2 ml/g
Total swelling volume acc. Houben Weyl (in DMF): ≥ 5.2 ml/g
100 - 200 mesh

Legal Information

Novabiochem is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Novabiochem® offers a wide range of linkers and derivatized resins for Fmoc solid-phase peptide synthesis with specialized protocols.

Protocols

Review various resins like Merrifield, trityl-based, and hydroxymethyl-functionalized for peptide immobilization for diverse applications.

Questions

  1. Hi. I need a resin for my synthesis where I can deprotect both Fmoc and Boc without cleaving the peptide from the resin. Is this resin suitable for this application? or do you have any suitable safety catch resin? Many thanks.

    1 answer
    1. Unfortunately, this material is not tested for suitability for any specific application. However, a brief search of the literature suggests that it may not be compatible for Boc peptides. Please see the links below to review Fmoc and Boc resin cleavage and protection information and products that may be helpful.

      Fmoc Resin Cleavage and Deprotection:
      https://www.sigmaaldrich.com/technical-documents/technical-article/chemistry-and-synthesis/peptide-synthesis/fmoc-cleavage-deprotection

      Boc Resin Cleavage and Deprotection
      https://www.sigmaaldrich.com/technical-documents/technical-article/chemistry-and-synthesis/peptide-synthesis/boc-resin-cleavage

      Please see the link below to review an additional publication that may be helpful:
      https://www.ncbi.nlm.nih.gov/pmc/articles/PMC11012524/

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service