Skip to Content
Merck
All Photos(1)

Key Documents

8.55059

Sigma-Aldrich

Merrifield resin LL (100-200 mesh)

Novabiochem®

Synonym(s):

Chloromethylpolystyrene-divinylbenzene (100-200 mesh)

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
€29.30
25 G
€103.00
100 G
€308.00

€29.30


Estimated to ship on18 May 2025


Request a Bulk Order

Select a Size

Change View
5 G
€29.30
25 G
€103.00
100 G
€308.00

About This Item

UNSPSC Code:
12352101
NACRES:
NA.22

€29.30


Estimated to ship on18 May 2025


Request a Bulk Order

Quality Level

product line

Novabiochem®

form

beads

reaction suitability

reaction type: Fmoc solid-phase peptide synthesis

manufacturer/tradename

Novabiochem®

application(s)

peptide synthesis

storage temp.

2-30°C

General description

Merrifield resin was the standard support for the synthesis of peptide acids by Boc SPPS [1,2]. In peptide chemistry, it is now really only used in the synthesis of small to medium sized peptides, as the benzylic ester resin linkage is not completely stable towards repetitive treatment with TFA. Attachment of the C-terminal residue is generally achieved by heating the resin in DMF with the appropriate amino acid cesium salt in the presence of KI [3]; although,Me4N salts [4], sodium salts[5] in THF with Bu4NF catalysis [6] and zinc salts in EtOH have also been used [7]. Cleavage is normally effected by treatment of resin with HF or TFMSA, or by hydrogenolysis [8]. Alcohols can be released using reducing agents such as DIBAL [6] or LiBH4 [9]. Methyl esters can be produced by transesterification with methoxide [10].

Associated Protocols and Technical Articles
Protocols for Loading of Peptide Synthesis Resins

Linkage

Replaces: 01-64-0008

Analysis Note

Color (visual): white to yellow to beige
Appearance of substance (visual): beads
Loading (determined by elemental analysis for sulfur after derivatization with KSAc): 0.40 - 1.00 mmol/g
Swelling Volume (in CH₂Cl₂): lot specific result
The polymer matrix is copoly (styrene-1 % DVB), 100 -200 mesh (75 - 150 µm)

Legal Information

Novabiochem is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service