Skip to Content
Merck

Skip To

8.55035

DFPE polystyrene

Novabiochem®

Synonym(s):

2-(3,5-Dimethoxy-4-formylphenoxy)ethyl polystyrene

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

NACRES:
NA.22
UNSPSC Code:
13111023
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

product line

Novabiochem®

form

beads

reaction suitability

reaction type: Fmoc solid-phase peptide synthesis

manufacturer/tradename

Novabiochem®

application(s)

peptide synthesis

functional group

aldehyde

storage temp.

2-8°C

General description

This resin is prepared under conditions optimized to minimize styrene formation during attachment of the linker to the functionalized ethylpolystyrene support. This high substitution, coupled with the lack of amide bonds and potentially acid-sensitive benzylic linkages, makes this support the preferred choice of SPOS of secondary sulfonamides and carboxamides. Applications of FMPE include aminofurazanopyrazines [1] and peptide agonists for human orphan receptor BRS-3 [2].

Associated Protocols and Technical Articles
Protocols for Loading of Peptide Synthesis Resins Literature references

Literature references

[1] B. Dörner & P. White in ′Peptides 1998, Proc. 25th European Peptide Symposium′, S. Bajusz & F. Hudecz (Eds), Budapest, Akadémiai Kiadó, 1999, pp. 90.
[2] M. Grimstrup & F. Zaragoza (2002) Eur. J. Org. Chem., 2953.
[3] K. J. Jensen, et al. (1998) J. Am. Chem. Soc., 120, 5441.
[4] M. del Fresno, et al. (1998) Tetrahedron Lett., 39, 2639.
[5] N. S. Gray, et al. (1997) Tetrahedron Lett., 38, 1161.
[6] J. Alsina, et al. (1999) Chem. Eur. J., 5, 2787.
[7] J. F. Tolborg & K. J. Jensen (2000) Chem. Commun., 147.
[8] A. E. Fenwick, et al. (2001) Bioorg. Med. Chem. Lett., 11, 195.
[9] J. Giovannoni, et al. (2001) Tetrahedron Lett., 42, 5389.
[10] J. T. Bork, et al. (2003) Org. Lett., 5, 117.
[11] C. P. Holmes, Paper ORGN 383 presented at the 213th National Meeting of the American Chemical Society, San Francisco, 1997.
[12] C. Pégurier, et al. (2000) Bioorg. Med. Chem., 8,163.
[13] T. F. Herpin, et al. (2000) J. Comb. Chem., 2, 513.

Analysis Note

Color (visual): white to yellow to beige
Appearance of substance (visual): beads
Loading (determined by elemental analysis of sulfur after derivatization with tosylhydrazide): 0.50 - 1.10 mmol/g
Swelling Volume (in DMF): lot specific result
The polymer matrix is copoly (styrene-1 % DVB), 100 -200 mesh

Other Notes

Replaces: 01-64-0360

Legal Information

Novabiochem is a registered trademark of Merck KGaA, Darmstadt, Germany

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
8.550688.55115327824
reaction suitability

reaction type: Fmoc solid-phase peptide synthesis

reaction suitability

reaction type: Boc solid-phase peptide synthesis

reaction suitability

reaction type: Boc solid-phase peptide synthesis

reaction suitability

-

functional group

aldehyde

functional group

alcohol

functional group

amine

functional group

-

product line

Novabiochem®

product line

Novabiochem®

product line

Novabiochem®

product line

-

Quality Level

200

Quality Level

200

Quality Level

200

Quality Level

100

application(s)

peptide synthesis

application(s)

peptide synthesis

application(s)

peptide synthesis

application(s)

-

form

beads

form

beads

form

beads

form

(Viscous Liquid or Solid or Chunk(s))


Storage Class

11 - Combustible Solids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters