Skip to Content
Merck

Skip To

8.53074

Boc-Asn(Trt)-OH

Novabiochem®

Synonym(s):

Boc-Asn(Trt)-OH, N-α-t.-Boc-β-trityl-L-asparagine

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
25 g
Check Cart for Availability
€ 863,00

About This Item

Empirical Formula (Hill Notation):
C28H30N2O5
CAS Number:
Molecular Weight:
474.55
UNSPSC Code:
12352209
NACRES:
NA.22
MDL number:

€ 863,00


Check Cart for Availability

Request a Custom Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

product line

Novabiochem®

assay

≥98% (TLC)

form

powder

reaction suitability

reaction type: Boc solid-phase peptide synthesis

manufacturer/tradename

Novabiochem®

application(s)

peptide synthesis

functional group

amide

storage temp.

15-25°C

SMILES string

N([C@@H](CC(=O)NC(c3ccccc3)(c2ccccc2)c1ccccc1)C(=O)O)C(=O)OC(C)(C)C

InChI

1S/C28H30N2O5/c1-27(2,3)35-26(34)29-23(25(32)33)19-24(31)30-28(20-13-7-4-8-14-20,21-15-9-5-10-16-21)22-17-11-6-12-18-22/h4-18,23H,19H2,1-3H3,(H,29,34)(H,30,31)(H,32,33)/t23-/m0/s1

InChI key

PYGOCFDOBSXROC-QHCPKHFHSA-N

General description

A trityl-protected derivative which increases the solubility of Asn during the coupling step in SPPS [1]. Its use gives higher yields and lower by-product formation. During the following coupling cycles the temporary side-chain protection is removed.

Associated Protocols and Technical Articles
Cleavage and Deprotection Protocols for Boc SPPS

Literature references

[1] P. Sieber, et al. (1991) Tetrahedron Lett., 32, 739.

Analysis Note

Colour (visual): white to slight yellow to beige
Appearance of substance (visual): powder
Identity (IR): passes test
Optical rotation α 25/D (c=1 in methanol): -21.5 - -16.5 °
Purity (TLC(011A)): ≥ 98 %
Purity (TLC(0811)): ≥ 98 %
Solubility (1 mmole in 2 ml DMF): clear solution

To see the solvent systems used for TLC of Novabiochem® products please click here.

Other Notes

Replaces: 04-12-0187

Legal Information

Novabiochem is a registered trademark of Merck KGaA, Darmstadt, Germany

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
8.520448.5211947672
form

powder

form

powder

form

powder

form

powder

functional group

amide

functional group

amide

functional group

amide

functional group

Fmoc

assay

≥98% (TLC)

assay

≥98% (TLC), ≥98.0% (acidimetric), ≥99.0% (HPLC)

assay

≥95.0% (acidimetric), ≥98.0% (HPLC), ≥98.0% (TLC)

assay

≥97.0%

product line

Novabiochem®

product line

Novabiochem®

product line

Novabiochem®

product line

-

storage temp.

15-25°C

storage temp.

−20°C (−15°C to −25°C)

storage temp.

15-25°C

storage temp.

2-8°C

Quality Level

200

Quality Level

400

Quality Level

200

Quality Level

100


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters