Skip to Content
Merck

Skip To

8.53053

Boc-Lys(Boc)-OH . DCHA

Novabiochem®

Synonym(s):

Boc-Lys(Boc)-OH . DCHA, N-α,ε-di-t.-Boc-L-lysine dicyclohexylammonium salt

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
25 g
Check Cart for Availability
€ 129,00

About This Item

Empirical Formula (Hill Notation):
C16H30N2O6 · xC12H23N
CAS Number:
Molecular Weight:
346.42
MDL number:
UNSPSC Code:
12352209
EC Index Number:
239-150-9
NACRES:
NA.22

€ 129,00


Check Cart for Availability

Request a Custom Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

product line

Novabiochem®

assay

≥98% (TLC)

form

powder

reaction suitability

reaction type: Boc solid-phase peptide synthesis

manufacturer/tradename

Novabiochem®

application(s)

peptide synthesis

functional group

amine

storage temp.

2-30°C

SMILES string

[N+H2](C2CCCCC2)C1CCCCC1.N([C@@H](CCCCNC(=O)OC(C)(C)C)C(=O)[O-])C(=O)OC(C)(C)C

InChI

1S/C16H30N2O6.C12H23N/c1-15(2,3)23-13(21)17-10-8-7-9-11(12(19)20)18-14(22)24-16(4,5)6;1-3-7-11(8-4-1)13-12-9-5-2-6-10-12/h11H,7-10H2,1-6H3,(H,17,21)(H,18,22)(H,19,20);11-13H,1-10H2/t11-;/m0./s1

InChI key

HRLHJTYAMCGERD-MERQFXBCSA-N

General description

Building block of introduction of N-terminal lysine amino-acid residues by Fmoc SPPS

Associated Protocols and Technical Articles
Cleavage and Deprotection Protocols for Boc SPPS

Analysis Note

Color (visual): white to slight yellow to beige
Appearance of substance (visual): powder
Identity (IR): passes test
Optical rotation α 25/D (c=1 in AcOH): -6.5 - -4.5 °
Purity (TLC(011A)): ≥ 98 %
Purity (TLC(0811)): ≥ 98 %

To see the solvent systems used for TLC of Novabiochem® products please click here.

Other Notes

Replaces: 04-12-0063

Legal Information

Novabiochem is a registered trademark of Merck KGaA, Darmstadt, Germany

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
8.530608.530848.53021
form

powder

form

powder

form

powder

form

powder

functional group

amine

functional group

-

functional group

-

functional group

hydroxyl

assay

≥98% (TLC)

assay

≥98% (TLC)

assay

≥98% (TLC)

assay

≥98% (TLC)

product line

Novabiochem®

product line

Novabiochem®

product line

Novabiochem®

product line

Novabiochem®

storage temp.

2-30°C

storage temp.

2-30°C

storage temp.

2-8°C

storage temp.

2-30°C

Quality Level

200

Quality Level

200

Quality Level

200

Quality Level

200


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters