Skip to Content
Merck

Skip To

8.53033

Boc-Cys(4-MeBzl)-OH

Novabiochem®

Synonym(s):

Boc-Cys(4-MeBzl)-OH, N-α-t.-Boc-S-p-methylbenzyl-L-cysteine

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
25 g
Check Cart for Availability
€ 180,00
€ 135,00
100 g
Check Cart for Availability
€ 642,00
€ 481,50

About This Item

Empirical Formula (Hill Notation):
C16H23NO4S
CAS Number:
Molecular Weight:
325.42
UNSPSC Code:
12352209
NACRES:
NA.22
MDL number:

€ 135,00

List Price€ 180,00Save 25%
Web-Only Promotion

Check Cart for Availability

Request a Bulk Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

product line

Novabiochem®

assay

≥98% (TLC), ≥98.5% (HPLC)

form

powder

reaction suitability

reaction type: Boc solid-phase peptide synthesis

manufacturer/tradename

Novabiochem®

mp

63-66 °C

application(s)

peptide synthesis

functional group

thiol

storage temp.

2-30°C

SMILES string

S(CC(NC(=O)OC(C)(C)C)C(=O)O)Cc1ccc(cc1)C

InChI

1S/C16H23NO4S/c1-11-5-7-12(8-6-11)9-22-10-13(14(18)19)17-15(20)21-16(2,3)4/h5-8,13H,9-10H2,1-4H3,(H,17,20)(H,18,19)

InChI key

CUNVVZWSABRKAL-UHFFFAOYSA-N

General description

Standard building block for introduction of cysteine amino-acid residues by Boc SPPS.

Associated Protocols and Technical Articles
Cleavage and Deprotection Protocols for Boc SPPS
Fmoc SPPS of Cysteine-Containing Peptides

Analysis Note

Color (visual): white to slight yellow to beige
Appearance of substance (visual): powder
Identity (IR): passes test
Purity (TLC(011A)): ≥ 98 %
Purity (TLC(0811)): ≥ 98 %
Assay (HPLC, area%): ≥ 98.5 % (a/a)
Solubility (1 mmole in 2 ml DMF): clearly soluble
Enantiomeric purity: ≥ 99.5 % (a/a)

To see the solvent systems used for TLC of Novabiochem® products please click here.

Other Notes

Replaces: 04-12-0134

Legal Information

Novabiochem is a registered trademark of Merck KGaA, Darmstadt, Germany

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
8.530508.530468.53049
form

powder

form

powder

form

powder

form

powder

functional group

thiol

functional group

thiol

functional group

thiol

functional group

thiol

assay

≥98% (TLC), ≥98.5% (HPLC)

assay

≥98% (TLC)

assay

≥98% (TLC)

assay

≥98% (TLC)

product line

Novabiochem®

product line

Novabiochem®

product line

Novabiochem®

product line

Novabiochem®

storage temp.

2-30°C

storage temp.

2-30°C

storage temp.

2-30°C

storage temp.

2-30°C

mp

63-66 °C

mp

-

mp

-

mp

-


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters