Skip to Content
Merck
All Photos(1)

Key Documents

8.52411

Sigma-Aldrich

Fmoc-Asn(Mmt)-OH

Novabiochem®

Synonym(s):

Fmoc-Asn(Mmt)-OH, N-α-Fmoc-N-β-p-methoxytrityl-L-asparagine

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
€68.30
5 G
€206.00
100 G
€2,700.00

€68.30


Estimated to ship on30 April 2025


Request a Bulk Order

Select a Size

Change View
1 G
€68.30
5 G
€206.00
100 G
€2,700.00

About This Item

Empirical Formula (Hill Notation):
C39H34N2O6
Molecular Weight:
626.70
UNSPSC Code:
12352209
NACRES:
NA.22

€68.30


Estimated to ship on30 April 2025


Request a Bulk Order

Quality Level

product line

Novabiochem®

form

powder

reaction suitability

reaction type: Fmoc solid-phase peptide synthesis

manufacturer/tradename

Novabiochem®

application(s)

peptide synthesis

functional group

amide

storage temp.

15-25°C

General description

A superior derivative to Fmoc-Asn(Trt)-OH for the synthesis of Asn-containing peptides by Fmoc SPPS. Fmoc-Asn(Mmt)-OH is more soluble in DMF than Fmoc-Asn(Trt)-OH, thereby facilitating coupling reactions at higher concentration. Cleavage of the Mmt group is rapid, even when the residue is located at the N-terminus of a peptide.

Associated Protocols and Technical Articles
Cleavage and Deprotection Protocols for Fmoc SPPS

Analysis Note

Color (visual): white to off white
Appearance of substance (visual): powder
Identity (IR): passes test
Solubility (1 mmole in 2 ml DMF): clearly soluble
Enantiomeric purity: ≥ 99.5 % (a/a)
Purity (HPLC): ≥ 99.0 % (a/a)
Purity (TLC(011A)): ≥ 98 %
Purity (TLC(0811)): ≥ 98 %
Acetate (IC): ≤ 0.1 %

To see the solvent systems used for TLC of Novabiochem® products please click here.

Legal Information

Novabiochem is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service