Skip to Content
Merck
All Photos(1)

Documents

8.52372

Sigma-Aldrich

Fmoc-selenomethionine

Novabiochem®

Synonym(s):

Fmoc-selenomethionine, N-Fmoc-L-amino-4-(methylselanyl)butanoic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H21NO4Se
CAS Number:
Molecular Weight:
418.35
UNSPSC Code:
12352209

Quality Level

product line

Novabiochem®

form

powder

reaction suitability

reaction type: Fmoc solid-phase peptide synthesis

manufacturer/tradename

Novabiochem®

application(s)

peptide synthesis

functional group

Fmoc

storage temp.

2-8°C

InChI

1S/C20H21NO4Se/c1-26-11-10-18(19(22)23)21-20(24)25-12-17-15-8-4-2-6-13(15)14-7-3-5-9-16(14)17/h2-9,17-18H,10-12H2,1H3,(H,21,24)(H,22,23)/t18-/m0/s1

InChI key

MOIJGLAOGAWHTN-SFHVURJKSA-N

General description

Fmoc-selenomethionine can be introduced under standard conditions. Any selenoxide formed during synthesis can be easily reduced back to selenide by treatment with β-mercaptoethanol. Introduction of selenium can help facilitate solid phase and solution structural determination and the study of peptide-protein interactions [1].

Associated Protocols and Technical Articles
Cleavage and Deprotection Protocols for Fmoc SPPS

Literature references

[1] L. Moroder (2005) J. Pept.Sci., 11, 187.

Legal Information

Novabiochem is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service