Skip to Content
Merck
All Photos(1)

Key Documents

8.52333

Sigma-Aldrich

(S)-N-Fmoc-alpha-4-n-pentenylalanine

Novabiochem®

Synonym(s):

(S)-N-Fmoc-alpha-4-n-pentenylalanine

Sign Into View Organizational & Contract Pricing

Select a Size

250 MG
€598.00

€598.00


Estimated to ship on17 May 2025


Request a Bulk Order

Select a Size

Change View
250 MG
€598.00

About This Item

Empirical Formula (Hill Notation):
C23H25NO4
CAS Number:
Molecular Weight:
379.45
MDL number:
UNSPSC Code:
12352209
NACRES:
NA.22

€598.00


Estimated to ship on17 May 2025


Request a Bulk Order

Quality Level

product line

Novabiochem®

form

solid

reaction suitability

reaction type: Fmoc solid-phase peptide synthesis

manufacturer/tradename

Novabiochem®

application(s)

peptide synthesis

functional group

alkene

storage temp.

15-25°C

SMILES string

N([C@](CCCC=C)(C)C(=O)O)C(=O)OCC1c2c(cccc2)c3c1cccc3

InChI

1S/C23H25NO4/c1-3-4-9-14-23(2,21(25)26)24-22(27)28-15-20-18-12-7-5-10-16(18)17-11-6-8-13-19(17)20/h3,5-8,10-13,20H,1,4,9,14-15H2,2H3,(H,24,27)(H,25,26)/t23-/m0/s1

InChI key

MRJFPZWLOJOINV-QHCPKHFHSA-N

General description

Building block for introduction of α-n-pentenyl-alanine peptides by Fmoc SPPS.

Analysis Note

Color (visual): white to slight yellow to beige
Appearance of substance (visual): powder
Identity (IR): passes test
Assay (HPLC, area%): ≥ 98.0 % (a/a)
Solubility (1 mmole in 2 ml DMF): clearly soluble

Legal Information

Novabiochem is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service