Skip to Content
Merck
All Photos(1)

Key Documents

8.52266

Sigma-Aldrich

Fmoc-hCys(Trt)-OH

Novabiochem®

Synonym(s):

Fmoc-hCys(Trt)-OH, Fmoc-S-trityl-L-homocysteine

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
€311.00
5 G
€1,230.00

€311.00


Estimated to ship on25 May 2025


Request a Bulk Order

Select a Size

Change View
1 G
€311.00
5 G
€1,230.00

About This Item

Empirical Formula (Hill Notation):
C38H33NO4S
CAS Number:
Molecular Weight:
599.74
MDL number:
UNSPSC Code:
12352209
NACRES:
NA.22

€311.00


Estimated to ship on25 May 2025


Request a Bulk Order

Quality Level

product line

Novabiochem®

Assay

≥95.0% (HPLC)
≥97% (TLC)

form

powder

reaction suitability

reaction type: Fmoc solid-phase peptide synthesis

manufacturer/tradename

Novabiochem®

application(s)

peptide synthesis

functional group

thiol

storage temp.

15-25°C

SMILES string

S(CC[C@H](NC(=O)OCC4c5c(cccc5)c6c4cccc6)C(=O)O)C(c3ccccc3)(c2ccccc2)c1ccccc1

InChI

1S/C38H33NO4S/c40-36(41)35(39-37(42)43-26-34-32-22-12-10-20-30(32)31-21-11-13-23-33(31)34)24-25-44-38(27-14-4-1-5-15-27,28-16-6-2-7-17-28)29-18-8-3-9-19-29/h1-23,34-35H,24-26H2,(H,39,42)(H,40,41)/t35-/m0/s1

InChI key

FKBGJLDYRSFHBT-DHUJRADRSA-N

General description

Fmoc-homoCys(Trt)-OH is a useful tool for the introduction of homocysteine during Fmoc SPPS. It can be used in exactly the same manner as Fmoc-Cys(Trt)-OH.

Associated Protocols and Technical Articles
Cleavage and Deprotection Protocols for Fmoc SPPS

Analysis Note

Colour (visual): white to slight yellow to beige
Appearance of substance (visual): powder
Identity (IR): passes test
Enantiomeric purity: ≥ 98.0 % (a/a)
Purity (TLC(011A)): ≥ 97 %
Assay (HPLC, area%): ≥ 95.0 % (a/a)
Solubility (1 mmole in 2 ml DMF): clearly soluble

To see the solvent systems used for TLC of Novabiochem® products please click here.

Legal Information

Novabiochem is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service