Skip to Content
Merck
All Photos(1)

Documents

8.52216

Sigma-Aldrich

Fmoc-Dpr(Boc-Aoa)-OH

Novabiochem®

Synonym(s):

Fmoc-Dpr(Boc-Aoa)-OH, N-α-Fmoc-N-β-(N-t.-Boc-amino-oxyacetyl)-L-diaminopropionic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C25H29N3O8
CAS Number:
Molecular Weight:
499.51
UNSPSC Code:
12352209
NACRES:
NA.22

Quality Level

product line

Novabiochem®

Assay

≥95.0% (acidimetric)
≥96.0% (HPLC)
≥98% (TLC)

form

powder

reaction suitability

reagent type: ligand

manufacturer/tradename

Novabiochem®

application(s)

peptide synthesis

functional group

Boc
Fmoc

storage temp.

2-30°C

InChI

1S/C25H29N3O8/c1-25(2,3)36-24(33)28-35-14-21(29)26-12-20(22(30)31)27-23(32)34-13-19-17-10-6-4-8-15(17)16-9-5-7-11-18(16)19/h4-11,19-20H,12-14H2,1-3H3,(H,26,29)(H,27,32)(H,28,33)(H,30,31)/t20-/m0/s1

InChI key

KHRQNEDYPNTPKW-FQEVSTJZSA-N

General description

An excellent derivative for the chemoselective ligation of unprotected peptides in aqueous media [1] via oxime formation. This derivative introduces a hydroxylamine functionality which can couple with aldehyde groups present in another peptide unit.

Associated Protocols and Technical Articles
Cleavage and Deprotection Protocols for Fmoc SPPS

Literature references

[1] F. Wahl & M. Mutter (1996) Tetrahedron Lett., 37, 6861.

Linkage

Replaces: 04-12-1185

Analysis Note

Color (visual): white to slight yellow to beige
Appearance of substance (visual): powder
Identity (IR): passes test
Enantiomeric purity: ≥ 99.5 % (a/a)
Purity (TLC(CMA2)): ≥ 98 %
Assay (HPLC, area%): ≥ 96.0 % (a/a)
Solubility (1 mmole in 2 ml DMF): clearly soluble
Assay (acidimetric): ≥ 95.0 %
Water (K. F.): ≤ 2.00 %

To see the solvent systems used for TLC of Novabiochem® products please click here.

Legal Information

Novabiochem is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service