Skip to Content
Merck
All Photos(1)

Key Documents

8.52118

Sigma-Aldrich

Fmoc-Asp(EDANS)-OH

Novabiochem®

Synonym(s):

Fmoc-Asp(EDANS)-OH

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C31H29N3O8S
CAS Number:
Molecular Weight:
603.64
MDL number:
UNSPSC Code:
12352209
Pricing and availability is not currently available.

Quality Level

product line

Novabiochem®

Assay

≥95.0% (HPLC)
≥98% (TLC)

form

powder

reaction suitability

reaction type: Fmoc solid-phase peptide synthesis

manufacturer/tradename

Novabiochem®

application(s)

peptide synthesis

functional group

Fmoc

storage temp.

15-25°C

SMILES string

[S](=O)(=O)(O)c1c2c(c(ccc2)NCCNC(=O)C[C@H](NC(=O)OCC3c4c(cccc4)c5c3cccc5)C(=O)O)ccc1

InChI

1S/C31H29N3O8S/c35-29(33-16-15-32-26-13-5-12-24-23(26)11-6-14-28(24)43(39,40)41)17-27(30(36)37)34-31(38)42-18-25-21-9-3-1-7-19(21)20-8-2-4-10-22(20)25/h1-14,25,27,32H,15-18H2,(H,33,35)(H,34,38)(H,36,37)(H,39,40,41)/t27-/m0/s1

InChI key

ZHXKVZRMHMFRBQ-MHZLTWQESA-N

General description

Fluorescence-labeled amino acid for preparing fluorescence-quenched peptide substrates by Fmoc SPPS [1]. Most frequently used in conjunction with Dabcyl quenching group.

Associated Protocols and Technical Articles
Cleavage and Deprotection Protocols for Fmoc SPPS
Chromogenic Labeling of Synthetic Peptides

Literature references

[1] L. L. Maggiora, et al. (1992) J. Med. Chem., 35, 3727.

Linkage

Replaces: 04-12-1288

Analysis Note

Color (visual): grey
Appearance of substance (visual): powder
Identity (IR): passes test
Enantiomeric purity: ≥ 99.5 % (a/a)
Purity (TLC(CMA1)): ≥ 98 %
Assay (HPLC, area%): ≥ 95.0 % (a/a)

To see the solvent systems used for TLC of Novabiochem® products please click here.

Legal Information

Novabiochem is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service