Skip to Content
Merck
All Photos(1)

Key Documents

8.52080

Sigma-Aldrich

Fmoc-Tyr(2-ClTrt)-OH

Novabiochem®

Synonym(s):

Fmoc-Tyr(2-ClTrt)-OH, N-α-Fmoc-O-2-chlorotrityl-L-tyrosine

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
€185.00
25 G
€731.00
100 G
€2,060.00

€185.00


Estimated to ship on14 May 2025


Request a Bulk Order

Select a Size

Change View
5 G
€185.00
25 G
€731.00
100 G
€2,060.00

About This Item

Empirical Formula (Hill Notation):
C43H34ClNO5
CAS Number:
Molecular Weight:
680.19
MDL number:
UNSPSC Code:
12352209
NACRES:
NA.22

€185.00


Estimated to ship on14 May 2025


Request a Bulk Order

Quality Level

product line

Novabiochem®

Assay

≥98% (TLC)
≥99.0% (HPLC)

form

powder

reaction suitability

reaction type: Fmoc solid-phase peptide synthesis

manufacturer/tradename

Novabiochem®

application(s)

peptide synthesis

functional group

hydroxyl

storage temp.

2-8°C

SMILES string

Clc1c(cccc1)C(Oc4ccc(cc4)C[C@H](NC(=O)OCC5c6c(cccc6)c7c5cccc7)C(=O)O)(c3ccccc3)c2ccccc2

InChI

1S/C43H34ClNO5/c44-39-22-12-11-21-38(39)43(30-13-3-1-4-14-30,31-15-5-2-6-16-31)50-32-25-23-29(24-26-32)27-40(41(46)47)45-42(48)49-28-37-35-19-9-7-17-33(35)34-18-8-10-20-36(34)37/h1-26,37,40H,27-28H2,(H,45,48)(H,46,47)/t40-/m0/s1

InChI key

XYAWVRLFZHVTNW-FAIXQHPJSA-N

General description

The side-chain ClTrt group can be removed with 1% TFA/ 5% TIS in DCM, enabling the side-chain hydroxyl group to be selectively modified whilst the derivative is attached to the solid support during Fmoc SPPS.

Associated Protocols and Technical Articles
Cleavage and Deprotection Protocols for Fmoc SPPS


Literature references

[1] K. Barlos, et al. (1998) J. Peptide Res., 51, 194.

Linkage

Replaces: 04-12-1184

Analysis Note

Color (visual): white to slight yellow to beige
Appearance of substance (visual): powder
Identity (IR): passes test
Enantiomeric purity: ≥ 99.5 % (a/a)
Purity (TLC(011A)): ≥ 98 %
Purity (TLC(0811)): ≥ 98 %
Assay (HPLC, area%): ≥ 99.0 % (a/a)
Solubility (1 mmole in 2 ml DMF): clearly soluble
Water (K. F.): ≤ 4.0 %

To see the solvent systems used for TLC of Novabiochem® products please click here.

Legal Information

Novabiochem is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Novabiochem® offers orthogonally protected amino acids for peptide synthesis, including cyclic and branched peptides.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service