Skip to Content
Merck
All Photos(1)

Key Documents

8.51222

Sigma-Aldrich

Oxyma Pure B

Novabiochem®

Synonym(s):

Oxyma Pure B

Sign Into View Organizational & Contract Pricing

Select a Size

25 G
€62.10
100 G
€209.00
500 G
€1,020.00

€62.10


Estimated to ship on28 May 2025


Request a Bulk Order

Select a Size

Change View
25 G
€62.10
100 G
€209.00
500 G
€1,020.00

About This Item

CAS Number:
Molecular Weight:
185.14
MDL number:
UNSPSC Code:
12352107
NACRES:
NA.22

€62.10


Estimated to ship on28 May 2025


Request a Bulk Order

Quality Level

product line

Novabiochem®

form

powder

reaction suitability

reaction type: Coupling Reactions

mp

137-142 °C

density

1.6 g/cm3

application(s)

peptide synthesis

storage temp.

2-8°C

InChI

1S/C6H7N3O4/c1-8-4(10)3(7-13)5(11)9(2)6(8)12/h13H,1-2H3

InChI key

BVSCIXRAKUKIET-UHFFFAOYSA-N

General description

Auxilliary nucleophile for peptide coupling which reduces enantiomerization.

Associated Protocols and Technical Articles
Guide to Selection of Coupling Reagents

Analysis Note

Color (visual): white to beige
Appearance of substance (visual): powder
Purity (HPLC): ≥ 99.0 % (a/a)
Identity (IR): passes test

Legal Information

Novabiochem is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service