Skip to Content
Merck
All Photos(1)

Key Documents

8.51083

Sigma-Aldrich

Boc-NH-(PEG)₂₇-COOH (88 atoms)

Novabiochem®

Synonym(s):

Boc-NH-(PEG)₂₇-COOH (88 atoms)

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
€1,520.00

€1,520.00


Estimated to ship on23 May 2025


Request a Bulk Order

Select a Size

Change View
1 G
€1,520.00

About This Item

Empirical Formula (Hill Notation):
C64H127NO32
Molecular Weight:
1422.68
UNSPSC Code:
12352106
NACRES:
NA.22

€1,520.00


Estimated to ship on23 May 2025


Request a Bulk Order

Quality Level

product line

Novabiochem®

Assay

≥95.0% (HPLC)
≥98% (TLC)

form

powder

reaction suitability

reaction type: Pegylations

manufacturer/tradename

Novabiochem®

application(s)

peptide synthesis

storage temp.

2-8°C

General description

This building block is prepared from highly purified PEG to ensure homogeneous products free from contaminating PEG oligomers.

Linkage

Replaces: 01-63-0151

Analysis Note

Color (visual): white to slight yellow to beige
Appearance of substance (visual): powder
Identity (LC-MS): 1420.8 - 1422.8
Purity (TLC(157B)): ≥ 98 %
Assay (HPLC, area%): ≥ 95.0 % (a/a)

To see the solvent systems used for TLC of Novabiochem® products please click here.

Legal Information

Novabiochem is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service