Skip to Content
Merck
All Photos(1)

Key Documents

8.51061

Sigma-Aldrich

2-Chlorotrityl chloride

Novabiochem®

Synonym(s):

2-Chlorotrityl chloride

Sign Into View Organizational & Contract Pricing

Select a Size

25 G
€174.00

€174.00


Estimated to ship on25 May 2025


Request a Bulk Order

Select a Size

Change View
25 G
€174.00

About This Item

Empirical Formula (Hill Notation):
C19H14Cl2
CAS Number:
Molecular Weight:
313.22
MDL number:
UNSPSC Code:
12352107
EC Index Number:
255-647-3
NACRES:
NA.22

€174.00


Estimated to ship on25 May 2025


Request a Bulk Order

Quality Level

product line

Novabiochem®

Assay

≥95.0% (HPLC)

form

powder

manufacturer/tradename

Novabiochem®

mp

133.0-137.0 °C

application(s)

peptide synthesis

storage temp.

2-30°C

SMILES string

ClC(c3c(cccc3)Cl)(c2ccccc2)c1ccccc1

InChI

1S/C19H14Cl2/c20-18-14-8-7-13-17(18)19(21,15-9-3-1-4-10-15)16-11-5-2-6-12-16/h1-14H

InChI key

JFLSOKIMYBSASW-UHFFFAOYSA-N

General description

Hyperacid-labile ester protecting group [1]. Introduced under extremely mild conditions by reacting together the acid, trityl chloride and DIPEA in DCM. Ideal for the protection of the C-terminal carboxylic acid group of C-terminal protected peptide fragments, monoprotection of diacids and as a general carboxylic acid protecting group in organic synthesis.


Associated Protocols and Technical Articles
Guide to Selection of Orthogonally-Protected Building Blocks for Fmoc SPPS

Literature references

[1] P. Athanassopoulos, et al. (1995) Tetrahedron Lett., 36, 5645.

Linkage

Replaces: 01-63-0045

Analysis Note

Color (visual): white to slight yellow to beige
Appearance of substance (visual): powder
Identity (IR): passes test
Melting range (lower value): ≥ 133 °C
Melting range (upper value): ≤ 137 °C
Assay (HPLC, area%): ≥ 95.0 % (a/a)

Legal Information

Novabiochem is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

CorrosionExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Met. Corr. 1 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

8A - Combustible, corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service