Skip to Content
Merck
All Photos(1)

Key Documents

8.48335

Sigma-Aldrich

Methanol

≥99.85% (GC), Bio-Methanol (Gas-to-Liquid) for synthesis

Synonym(s):

Methanol, MeOH, Hydroxymethane, Methyl alcohol, Carbinol, Methyl alcohol

Sign Into View Organizational & Contract Pricing

Select a Size

50 L
€476.00

€476.00


Estimated to ship on12 April 2025



Select a Size

Change View
50 L
€476.00

About This Item

Linear Formula:
CH3OH
CAS Number:
Molecular Weight:
32.04
Beilstein:
1098229
MDL number:
UNSPSC Code:
12352104
EC Index Number:
200-659-6
NACRES:
NA.22
Assay:
≥99.85% (GC)
bp:
64.7 °C (lit.)
vapor pressure:
128 hPa ( 20 °C)
410 mmHg ( 50 °C)
97.68 mmHg ( 20 °C)

€476.00


Estimated to ship on12 April 2025


Product Name

Bio-Methanol (Gas-to-Liquid), for synthesis

vapor density

1.11 (vs air)

Quality Level

vapor pressure

128 hPa ( 20 °C)
410 mmHg ( 50 °C)
97.68 mmHg ( 20 °C)

Assay

≥99.85% (GC)

form

liquid

autoignition temp.

725 °F

potency

5628 mg/kg LD50, oral (Rat)
17100 mg/kg LD50, skin (Rabbit)

expl. lim.

36 %

refractive index

n20/D 1.329 (lit.)

kinematic viscosity

0.54-0.59 cSt(20 °C)

bp

64.7 °C (lit.)

mp

−98 °C (lit.)

transition temp

flash point 9.7 °C

density

0.791 g/mL at 25 °C (lit.)

storage temp.

2-30°C

SMILES string

CO

InChI

1S/CH4O/c1-2/h2H,1H3

InChI key

OKKJLVBELUTLKV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Analysis Note

Appearance (visual): clear
Assay (GC, area%): ≥ 99.85 % (a/a)
Colour number (Hazen): ≤ 5
Density (d 20 °C/ 4 °C): ≥ 0.790
Water (K. F.): ≤ 0.10 %
Non-volatile matter: ≤ 0.0008 %
Acidity (Acetic acid): ≤ 0.003 %
Evaporation Number: 1.9

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

49.5 °F - closed cup

Flash Point(C)

9.7 °C - closed cup


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH Annex XVII (Restriction List)

CAS No.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service