Skip to Content
Merck
All Photos(1)

Key Documents

8.43967

Sigma-Aldrich

1,2,3-Tribromopropane

for synthesis

Synonym(s):

1,2,3-Tribromopropane

Sign Into View Organizational & Contract Pricing

Select a Size

50 ML
€125.00
250 ML
€535.00

€125.00


Estimated to ship on24 May 2025


Request a Bulk Order

Select a Size

Change View
50 ML
€125.00
250 ML
€535.00

About This Item

Empirical Formula (Hill Notation):
C3H5Br3
CAS Number:
Molecular Weight:
280.78
MDL number:
UNSPSC Code:
12352100
EC Index Number:
202-478-8
NACRES:
NA.22

€125.00


Estimated to ship on24 May 2025


Request a Bulk Order

Quality Level

form

liquid

bp

220 °C/1013 hPa

mp

16-17 °C

transition temp

flash point 93 °C

density

2.398 g/cm3 at 25 °C

storage temp.

2-30°C

SMILES string

BrC(CBr)CBr

InChI

1S/C3H5Br3/c4-1-3(6)2-5/h3H,1-2H2

InChI key

FHCLGDLYRUPKAM-UHFFFAOYSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

199.4 °F - closed cup

Flash Point(C)

93 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service