Skip to Content
Merck
All Photos(2)

Key Documents

8.43949

Sigma-Aldrich

Cyclopentyl methyl ether

(stabilised) for synthesis

Synonym(s):

CPME

Sign Into View Organizational & Contract Pricing

Select a Size

250 ML
€48.10
1 L
€131.00

€48.10


Estimated to ship on24 May 2025


Request a Bulk Order

Select a Size

Change View
250 ML
€48.10
1 L
€131.00

About This Item

Empirical Formula (Hill Notation):
C6H12O
CAS Number:
Molecular Weight:
100.16
MDL number:
UNSPSC Code:
12352112
NACRES:
NA.22
grade:
for synthesis
synthesis grade
bp:
106 °C/760 mmHg
vapor pressure:
59.9 hPa ( 25 °C)

€48.10


Estimated to ship on24 May 2025


Request a Bulk Order

grade

for synthesis
synthesis grade

Quality Level

vapor pressure

59.9 hPa ( 25 °C)

Assay

≥99% (GC)

form

liquid

potency

200-2000 mg/kg LD50, oral (Rat)
>2000 mg/kg LD50, skin (Rat)

dilution

(for synthesis)

bp

106 °C/760 mmHg

mp

-140 °C

transition temp

flash point -1 °C

density

0.86 g/mL at 25 °C

storage temp.

2-30°C

SMILES string

COC1CCCC1

InChI

1S/C6H12O/c1-7-6-4-2-3-5-6/h6H,2-5H2,1H3

InChI key

SKTCDJAMAYNROS-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Analysis Note

Assay (GC, area%): ≥ 99.0 % (a/a)
Density (d 20 °C/ 4 °C): 0.862 - 0.864
Water (K. F.): ≤ 0.50 %
Peroxide (as H₂O₂): ≤ 0.05 %
Identity (IR): passes test

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Flam. Liq. 2 - Skin Irrit. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

37.4 °F - closed cup

Flash Point(C)

< 3 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service