Skip to Content
Merck
All Photos(1)

Documents

8.43866

Sigma-Aldrich

Polyvinyl alcohol, fully hydrolized

(Mw approx. 60000) for synthesis

Synonym(s):

Poly(vinyl alcohol), PVA, PVOH

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[-CH2CHOH-]n
CAS Number:
MDL number:
UNSPSC Code:
13111042
NACRES:
NA.22

Quality Level

form

solid

potency

>2000 mg/kg LD50, oral (Rat)

mp

160-240 °C

bulk density

400‑670 kg/m3

storage temp.

2-30°C

InChI

1S/C2H4O/c1-2-3/h2-3H,1H2

InChI key

IMROMDMJAWUWLK-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Analysis Note

Degree of hydrolysis: ≥ 98.0 %
dyn.Viscosity, 20°C (4 %; water): 9.0 - 11.0 mPa∙s
Loss on drying (110 °C 2h): ≤ 5.0 %
Ash: ≤ 1.0 %
Ester value: 15 - 25

Storage Class Code

11 - Combustible Solids

WGK

WGK 1


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH Annex XVII (Restriction List)

CAS No.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service