Skip to Content
Merck
All Photos(1)

Key Documents

8.43836

Sigma-Aldrich

Nickel boride

≥98% (ICP)

Synonym(s):

Nickel boride

Sign Into View Organizational & Contract Pricing

Select a Size

10 G
€120.00

€120.00


Estimated to ship on23 May 2025


Request a Bulk Order

Select a Size

Change View
10 G
€120.00

About This Item

Empirical Formula (Hill Notation):
BNi2
CAS Number:
Molecular Weight:
128.20
MDL number:
UNSPSC Code:
12352302
EC Index Number:
234-494-6
NACRES:
NA.22

€120.00


Estimated to ship on23 May 2025


Request a Bulk Order

Quality Level

Assay

≥98% (ICP)

form

powder

storage temp.

2-30°C

SMILES string

[Ni]#[B]=[Ni]

InChI

1S/B.2Ni

InChI key

WRLJWIVBUPYRTE-UHFFFAOYSA-N

General description

for synthesis

Analysis Note

Assay (ICP OES): ≥ 98.0 %

Signal Word

Danger

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 1A Inhalation - Skin Sens. 1 - STOT RE 1

Storage Class Code

6.1C - Combustible, acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH Annex XVII (Restriction List)

CAS No.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service