Skip to Content
Merck
All Photos(1)

Key Documents

8.43833

Sigma-Aldrich

1-Chloro-3-methoxypropane

for synthesis

Synonym(s):

1-Chloro-3-methoxypropane

Sign Into View Organizational & Contract Pricing

Select a Size

25 ML
€97.10

€97.10


Estimated to ship on28 May 2025


Request a Bulk Order

Select a Size

Change View
25 ML
€97.10

About This Item

Empirical Formula (Hill Notation):
C4H9ClO
CAS Number:
Molecular Weight:
108.57
MDL number:
UNSPSC Code:
12352112
EC Index Number:
252-919-3
NACRES:
NA.22

€97.10


Estimated to ship on28 May 2025


Request a Bulk Order

vapor pressure

25 hPa ( 20 °C)

Quality Level

Assay

≥99.0% (GC)

form

liquid

autoignition temp.

205 °C (DIN 51794)

potency

>2000 mg/kg LD50, oral (Rat)

mp

<-70 °C

transition temp

flash point 18 °C

density

0.999 g/cm3 at 20 °C

storage temp.

2-30°C

SMILES string

ClCCCOC

InChI

1S/C4H9ClO/c1-6-4-2-3-5/h2-4H2,1H3

InChI key

BQLHMMQUVJCTAN-UHFFFAOYSA-N

Analysis Note

Assay (GC, area%): ≥ 99.0 % (a/a)
Density (d 20 °C/ 4 °C): 0.997 - 1.001
Identity (IR): passes test

Pictograms

Flame

Signal Word

Danger

Hazard Statements

Hazard Classifications

Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

64.4 °F

Flash Point(C)

18 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service