Skip to Content
Merck
All Photos(1)

Key Documents

8.42857

Sigma-Aldrich

4,4′-Methylenebis(2,6-di-tert-butylphenol)

for synthesis

Synonym(s):

4,4′-Methylenebis(2,6-di-tert-butylphenol)

Sign Into View Organizational & Contract Pricing

Select a Size

10 G
€26.60
100 G
€41.30

€26.60


Estimated to ship on19 May 2025


Request a Bulk Order

Select a Size

Change View
10 G
€26.60
100 G
€41.30

About This Item

Empirical Formula (Hill Notation):
C29H44O2
CAS Number:
Molecular Weight:
424.66
MDL number:
UNSPSC Code:
12352104
EC Index Number:
204-279-1
NACRES:
NA.22

€26.60


Estimated to ship on19 May 2025


Request a Bulk Order

vapor pressure

<0.1 hPa ( 25 °C)

Quality Level

Assay

≥98.0% (GC)

form

crystals

potency

>5000 mg/kg LD50, oral (Rat)
>2000 mg/kg LD50, skin (Rabbit)

bp

217 °C/1 hPa

mp

153-156 °C

transition temp

flash point 222 °C

solubility

<0.0001 g/L

density

0.99 g/cm3 at 20 °C

storage temp.

2-30°C

SMILES string

Oc1c(cc(cc1C(C)(C)C)Cc2cc(c(c(c2)C(C)(C)C)O)C(C)(C)C)C(C)(C)C

InChI

1S/C29H44O2/c1-26(2,3)20-14-18(15-21(24(20)30)27(4,5)6)13-19-16-22(28(7,8)9)25(31)23(17-19)29(10,11)12/h14-17,30-31H,13H2,1-12H3

InChI key

MDWVSAYEQPLWMX-UHFFFAOYSA-N

Application


  • Improvement in biolubricant performance: The use of 4,4′-Methylenebis(2,6-di-tert-butylphenol) as an antioxidant additive significantly enhances the oxidative stability and performance of new biolubricant formulations, marking it as an effective solution in sustainable lubrication technology (Quinchia et al., 2011).

  • Study of bioaccumulation potential in soils: Research utilized 4,4′-Methylenebis(2,6-di-tert-butylphenol) to investigate its bioaccumulation potential in field-collected soils, demonstrating its environmental interaction and mobility which are crucial for risk assessments in ecological contexts (Princz et al., 2014).

  • Antioxidant activity and intramolecular interactions: A study explored the antioxidant activity of o-bisphenols like 4,4′-Methylenebis(2,6-di-tert-butylphenol), focusing on intramolecular hydrogen bonding and its role in enhancing radical scavenging efficiency, pivotal for designing more effective antioxidants (Amorati et al., 2003).

Analysis Note

Assay (GC, area%): ≥ 98.0 % (a/a)
Melting range (lower value): ≥ 153 °C
Melting range (upper value): ≤ 156 °C
Identity (IR): passes test

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Chronic 4

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

431.6 °F

Flash Point(C)

222 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service