Skip to Content
Merck
All Photos(1)

Key Documents

8.42116

Sigma-Aldrich

1,3-Dimethylbarbituric acid

for synthesis

Synonym(s):

1,3-Dimethylbarbituric acid

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
€52.10
100 G
€150.00

€52.10


Estimated to ship on19 May 2025


Request a Bulk Order

Select a Size

Change View
5 G
€52.10
100 G
€150.00

About This Item

CAS Number:
Molecular Weight:
156.14
MDL number:
UNSPSC Code:
12352115
EC Index Number:
212-211-7
NACRES:
NA.22

€52.10


Estimated to ship on19 May 2025


Request a Bulk Order

Quality Level

form

solid

potency

1780 mg/kg LD50, oral (Rat)

pH

2.5 (20 °C, 100 g/L in H2O, slurry)

mp

120-124 °C

solubility

60 g/L

bulk density

500 kg/m3

storage temp.

2-30°C

SMILES string

N1(C(=O)N(C(=O)CC1=O)C)C

InChI

1S/C6H8N2O3/c1-7-4(9)3-5(10)8(2)6(7)11/h3H2,1-2H3

InChI key

VVSASNKOFCZVES-UHFFFAOYSA-N

General description

for synthesis

Application

1,3-Dimethylbarbituric acid for synthesis. CAS 769-42-6, pH 2.5 (100 g/l, H₂O, 20 °C) (slurry).

Analysis Note

Assay (acidimetric)≥ 98.0 %Melting range (lower value)≥ 120 °CMelting range (upper value)≤ 124 °CIdentity (IR)passes test

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service