Skip to Content
Merck
All Photos(2)

Key Documents

8.42016

Sigma-Aldrich

(3-Chloropropyl)trichlorosilane

for synthesis

Synonym(s):

(3-Chloropropyl)trichlorosilane

Sign Into View Organizational & Contract Pricing

Select a Size

5 ML
€54.60
25 ML
€170.00

€54.60


0 left in stock

Request a Bulk Order

Select a Size

Change View
5 ML
€54.60
25 ML
€170.00

About This Item

Empirical Formula (Hill Notation):
C3H6Cl4Si
CAS Number:
Molecular Weight:
211.98
MDL number:
UNSPSC Code:
12352103
EC Index Number:
219-844-8
NACRES:
NA.22

€54.60


0 left in stock

Request a Bulk Order

Quality Level

Assay

≥98.0% (GC)

form

liquid

bp

81 °C/27 hPa

transition temp

flash point 80 °C

density

1.360 g/cm3 at 20 °C

storage temp.

2-30°C

SMILES string

[Si](Cl)(Cl)(Cl)CCCCl

InChI

1S/C3H6Cl4Si/c4-2-1-3-8(5,6)7/h1-3H2

InChI key

OOXSLJBUMMHDKW-UHFFFAOYSA-N

Analysis Note

Assay (GC, area%): ≥ 98.0 % (a/a)
Density (d 20 °C/ 4 °C): 1.358 - 1.362
Identity (IR): passes test

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Skin Corr. 1B

Supplementary Hazards

Storage Class Code

8A - Combustible, corrosive hazardous materials

WGK

WGK 1

Flash Point(F)

176.0 °F

Flash Point(C)

80 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service