Skip to Content
Merck
All Photos(1)

Key Documents

8.41778

Sigma-Aldrich

Butyrophenone

for synthesis

Synonym(s):

Butyrophenone

Sign Into View Organizational & Contract Pricing

Select a Size

25 ML
€48.10
250 ML
€318.00

€48.10


Estimated to ship on21 May 2025


Request a Bulk Order

Select a Size

Change View
25 ML
€48.10
250 ML
€318.00

About This Item

Empirical Formula (Hill Notation):
C10H12O
CAS Number:
Molecular Weight:
148.20
MDL number:
UNSPSC Code:
12352115
EC Index Number:
207-799-7
NACRES:
NA.22

€48.10


Estimated to ship on21 May 2025


Request a Bulk Order

Quality Level

Assay

≥98.0% (GC)

form

liquid

bp

220-222 °C/1013 hPa

mp

11-13 °C

transition temp

flash point 88 °C

density

0.988 g/cm3

storage temp.

2-30°C

SMILES string

O=C(CCC)c1ccccc1

InChI

1S/C10H12O/c1-2-6-10(11)9-7-4-3-5-8-9/h3-5,7-8H,2,6H2,1H3

InChI key

FFSAXUULYPJSKH-UHFFFAOYSA-N

Analysis Note

Assay (GC, area%): ≥ 98.0 % (a/a)
Identity (IR): passes test
Due to its specific melting range the product may be solid, liquid, a solidified melt or a supercooled melt.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

192.2 °F - closed cup

Flash Point(C)

89 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service