Skip to Content
Merck
All Photos(1)

Key Documents

8.41751

Sigma-Aldrich

Diethyl dicarbonate

for synthesis

Synonym(s):

Diethyl dicarbonate

Sign Into View Organizational & Contract Pricing

Select a Size

5 ML
€38.00
25 ML
€115.00

€38.00


Estimated to ship on24 May 2025


Request a Bulk Order

Select a Size

Change View
5 ML
€38.00
25 ML
€115.00

About This Item

Empirical Formula (Hill Notation):
C6H10O5
CAS Number:
Molecular Weight:
162.14
MDL number:
UNSPSC Code:
12352108
EC Index Number:
216-542-8
NACRES:
NA.22

€38.00


Estimated to ship on24 May 2025


Request a Bulk Order

Quality Level

Assay

≥98.0% (acidimetric)

form

liquid

potency

850 mg/kg LD50, oral (Rat)

bp

93-94 °C/24 hPa

transition temp

flash point 69 °C

density

1.12 g/cm3 at 20 °C

storage temp.

2-8°C

SMILES string

O(CC)C(=O)OC(=O)OCC

InChI

1S/C6H10O5/c1-3-9-5(7)11-6(8)10-4-2/h3-4H2,1-2H3

InChI key

FFYPMLJYZAEMQB-UHFFFAOYSA-N

Analysis Note

Assay (acidimetric): ≥ 98.0 %
Density (d 20 °C/ 4 °C): 1.118 - 1.122
Identity (IR): passes test

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

156.2 °F - closed cup

Flash Point(C)

69 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service