Skip to Content
Merck
All Photos(1)

Key Documents

8.41692

Sigma-Aldrich

N,N,N′,N′′,N′′-Pentamethyldiethylenetriamine

for synthesis

Synonym(s):

N,N,N′,N′′,N′′-Pentamethyldiethylenetriamine

Sign Into View Organizational & Contract Pricing

Select a Size

100 ML
€115.00
500 ML
€458.00

€115.00


Estimated to ship on25 May 2025


Request a Bulk Order

Select a Size

Change View
100 ML
€115.00
500 ML
€458.00

About This Item

Empirical Formula (Hill Notation):
C9H23N3
CAS Number:
Molecular Weight:
173.30
MDL number:
UNSPSC Code:
12352116
EC Index Number:
221-201-1
NACRES:
NA.22

€115.00


Estimated to ship on25 May 2025


Request a Bulk Order

vapor pressure

0.3 hPa ( 20 °C)

Quality Level

Assay

≥98.0% (GC)

form

liquid

autoignition temp.

155 °C

potency

1330 mg/kg LD50, oral (Rat)
230 mg/kg LD50, skin (Rabbit)

expl. lim.

1.1-5.7 % (v/v)

pH

11.9 (20 °C, 100 g/L in H2O)

mp

<-18 °C

transition temp

flash point 76 °C

density

0.828 g/cm3 at 20 °C

storage temp.

2-30°C

SMILES string

N(CCN(C)C)(CCN(C)C)C

InChI

1S/C9H23N3/c1-10(2)6-8-12(5)9-7-11(3)4/h6-9H2,1-5H3

InChI key

UKODFQOELJFMII-UHFFFAOYSA-N

Analysis Note

Assay (GC, area%): ≥ 98.0 % (a/a)
Identity (IR): passes test

Pictograms

CorrosionSkull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 4 Oral - Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

6.1A - Combustible, acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 2

Flash Point(F)

168.8 °F - closed cup

Flash Point(C)

76 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service