Skip to Content
Merck
All Photos(1)

Key Documents

8.41522

Sigma-Aldrich

1-Ethynylcyclohexanol

for synthesis

Synonym(s):

1-Ethynylcyclohexanol

Sign Into View Organizational & Contract Pricing

Select a Size

100 ML
€59.90
25 L
€6,250.00

€59.90


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
100 ML
€59.90
25 L
€6,250.00

About This Item

Empirical Formula (Hill Notation):
C8H12O
CAS Number:
Molecular Weight:
124.18
MDL number:
UNSPSC Code:
12352104
EC Index Number:
201-100-9
NACRES:
NA.22

€59.90


Check Cart for Availability

Request a Bulk Order

vapor pressure

1 hPa ( 50 °C)

Quality Level

Assay

≥98.0% (GC)

form

solid

autoignition temp.

365 °C

potency

583 mg/kg LD50, oral (Rat)
973 mg/kg LD50, skin (Rabbit)

expl. lim.

1.3-8.7 % (v/v)

pH

7 (20 °C, 1 g/L in H2O)

bp

180 °C/1013 hPa

mp

30-32 °C

transition temp

flash point 73 °C

solubility

10 g/L

density

0.976 g/cm3 at 20 °C

storage temp.

2-30°C

SMILES string

OC1(CCCCC1)C#C

InChI

1S/C8H12O/c1-2-8(9)6-4-3-5-7-8/h1,9H,3-7H2

InChI key

QYLFHLNFIHBCPR-UHFFFAOYSA-N

Analysis Note

Assay (GC, area%): ≥ 98.0 % (a/a)
Density (d 20 °C/ 4 °C): 0.972 - 0.979
Identity (IR): passes test
Due to its specific melting range the product may be solid, liquid, a solidified melt or a supercooled melt.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

6.1C - Combustible, acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 1

Flash Point(F)

163.4 °F - closed cup

Flash Point(C)

73 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service