Skip to Content
Merck
All Photos(2)

Key Documents

8.41072

Sigma-Aldrich

2′-Chloroacetophenone

for synthesis

Synonym(s):

2′-Chloroacetophenone, 1-(2-Chlorophenyl)ethanone

Sign Into View Organizational & Contract Pricing

Select a Size

50 ML
€117.00

€117.00


Estimated to ship on27 May 2025


Request a Bulk Order

Select a Size

Change View
50 ML
€117.00

About This Item

Empirical Formula (Hill Notation):
C8H7ClO
CAS Number:
Molecular Weight:
154.59
MDL number:
UNSPSC Code:
12352115
EC Index Number:
218-397-6
NACRES:
NA.22

€117.00


Estimated to ship on27 May 2025


Request a Bulk Order

Quality Level

Assay

≥98.0% (GC)

form

liquid

potency

1820 mg/kg LD50, oral (Rat)

transition temp

flash point 88 °C

density

1.189 g/cm3 at 20 °C

storage temp.

2-30°C

SMILES string

Clc1c(cccc1)C(=O)C

InChI

1S/C8H7ClO/c1-6(10)7-4-2-3-5-8(7)9/h2-5H,1H3

InChI key

ZDOYHCIRUPHUHN-UHFFFAOYSA-N

Analysis Note

Assay (GC, area%): ≥ 98.0 % (a/a)
Density (d 20 °C/ 4 °C): 1.188 - 1.192
Identity (IR): passes test

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1A - Combustible, acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

190.4 °F

Flash Point(C)

88 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service