Skip to Content
Merck
All Photos(1)

Key Documents

8.41025

Sigma-Aldrich

Vanillic acid

for synthesis

Synonym(s):

Vanillic acid, 4-Hydroxy-3-methoxybenzoic acid

Sign Into View Organizational & Contract Pricing

Select a Size

50 G
€103.00

€103.00


Estimated to ship on28 May 2025


Request a Bulk Order

Select a Size

Change View
50 G
€103.00

About This Item

Empirical Formula (Hill Notation):
C8H8O4
CAS Number:
Molecular Weight:
168.15
MDL number:
UNSPSC Code:
12352106
EC Index Number:
204-466-8
NACRES:
NA.22

€103.00


Estimated to ship on28 May 2025


Request a Bulk Order

Quality Level

Assay

≥98.0% (acidimetric)

form

powder

mp

208-210 °C

storage temp.

2-30°C

SMILES string

O(C)c1c(ccc(c1)C(=O)O)O

InChI

1S/C8H8O4/c1-12-7-4-5(8(10)11)2-3-6(7)9/h2-4,9H,1H3,(H,10,11)

InChI key

WKOLLVMJNQIZCI-UHFFFAOYSA-N

Analysis Note

Assay (acidimetric): ≥ 98.0 %
Melting range (lower value): ≥ 208 °C
Melting range (upper value): ≤ 212 °C
Identity (IR): passes test

Storage Class Code

11 - Combustible Solids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 2

1 of 2

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service