Skip to Content
Merck
All Photos(1)

Key Documents

8.40144

Sigma-Aldrich

Diethylene glycol divinyl ether

for synthesis

Synonym(s):

Diethylene glycol divinyl ether

Sign Into View Organizational & Contract Pricing

Select a Size

50 ML
€177.00

€177.00


Estimated to ship on28 May 2025


Request a Bulk Order

Select a Size

Change View
50 ML
€177.00

About This Item

Empirical Formula (Hill Notation):
C8H14O3
CAS Number:
Molecular Weight:
158.19
MDL number:
UNSPSC Code:
12352112
EC Index Number:
212-133-3
NACRES:
NA.22

€177.00


Estimated to ship on28 May 2025


Request a Bulk Order

Quality Level

Assay

≥98.0% (GC)

form

liquid

density

0.97 g/cm3

storage temp.

2-30°C

SMILES string

O(CCOC=C)CCOC=C

InChI

1S/C8H14O3/c1-3-9-5-7-11-8-6-10-4-2/h3-4H,1-2,5-8H2

InChI key

SAMJGBVVQUEMGC-UHFFFAOYSA-N

Analysis Note

Assay (GC, area%): ≥ 98.0 % (a/a)
Density (d 20 °C/ 4 °C): 0.968 - 0.972
Identity (IR): passes test

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

159.8 °F - closed cup

Flash Point(C)

71 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

  1. Is this stabilized by KOH or any other stabilizers?

    1 answer
    1. We cannot confirm the presence of a stabilizer in this product.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service