Skip to Content
Merck
All Photos(1)

Key Documents

8.40122

Sigma-Aldrich

Span® 65

for synthesis

Synonym(s):

Span® 65, Sorbitan tristearate, Sorbitan trioctadecanoate

Sign Into View Organizational & Contract Pricing

Select a Size

100 G
€36.30
500 G
€71.60

€36.30


Estimated to ship on28 May 2025


Request a Bulk Order

Select a Size

Change View
100 G
€36.30
500 G
€71.60

About This Item

Empirical Formula (Hill Notation):
C60H114O8
CAS Number:
Molecular Weight:
963.54
MDL number:
UNSPSC Code:
12352108
EC Index Number:
247-891-4
NACRES:
NA.22

€36.30


Estimated to ship on28 May 2025


Request a Bulk Order

vapor pressure

<1.4 hPa ( 20 °C)

Quality Level

form

powder

potency

>15900 mg/kg LD50, oral (Rat)

bp

>100 °C/1013 hPa

mp

53 °C

transition temp

flash point >149 °C

density

0.98 g/cm3 at 25 °C

storage temp.

2-30°C

SMILES string

O1[C@@H]([C@H]([C@@H](C1)O)OC(=O)CCCCCCCCCCCCCCCCC)C(OC(=O)CCCCCCCCCCCCCCCCC)COC(=O)CCCCCCCCCCCCCCCCC

InChI

1S/C60H114O8/c1-4-7-10-13-16-19-22-25-28-31-34-37-40-43-46-49-56(62)65-53-55(67-57(63)50-47-44-41-38-35-32-29-26-23-20-17-14-11-8-5-2)60-59(54(61)52-66-60)68-58(64)51-48-45-42-39-36-33-30-27-24-21-18-15-12-9-6-3/h54-55,59-61H,4-53H2,1-3H3/t54-,55?,59+,60-/m1/s1

InChI key

IJCWFDPJFXGQBN-BIFNRIDTSA-N

Application

Span® 65 for synthesis. CAS 26658-19-5, molar mass 963.55 g/mol.

Analysis Note

Hydroxyl value: 66 - 80
Saponification value: 176 - 188
Identity (IR): passes test

Legal Information

Span is a registered trademark of Croda International PLC

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

302.0 °F - closed cup

Flash Point(C)

150.00 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service