Skip to Content
Merck
All Photos(2)

Key Documents

8.40072

Sigma-Aldrich

Thioflavine T (C.I.No.49005)

≥73% sulfur basis

Synonym(s):

Thioflavine T (C.I.No.49005)

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
€81.20

€81.20


Estimated to ship on24 May 2025


Request a Bulk Order

Select a Size

Change View
5 G
€81.20

About This Item

Empirical Formula (Hill Notation):
C17H19ClN2S
CAS Number:
Molecular Weight:
318.86
MDL number:
UNSPSC Code:
12171501
EC Index Number:
219-228-9
NACRES:
NA.22

€81.20


Estimated to ship on24 May 2025


Request a Bulk Order

Quality Level

Assay

≥73% sulfur basis

form

powder

storage temp.

2-30°C

SMILES string

S1c2c(ccc(c2)C)N(C1=C3C=CC(=[N+](C)C)C=C3)C.[Cl-]

InChI

1S/C17H19N2S.ClH/c1-12-5-10-15-16(11-12)20-17(19(15)4)13-6-8-14(9-7-13)18(2)3;/h5-11H,1-4H3;1H/q+1;/p-1

InChI key

JADVWWSKYZXRGX-UHFFFAOYSA-M

General description

for synthesis

Analysis Note

Assay (of S): ≥ 73.0 %
Identity (IR): passes test

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1 - Skin Sens. 1

Storage Class Code

6.1C - Combustible, acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service