Skip to Content
Merck
All Photos(1)

Documents

8.22340

Sigma-Aldrich

Phosphorus pentachloride

for synthesis

Synonym(s):

Phosphorus pentachloride, Phosphorus(V) chloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
Cl5P
CAS Number:
Molecular Weight:
208.24
MDL number:
UNSPSC Code:
12352307
EC Index Number:
233-060-3
NACRES:
NA.22

vapor pressure

0.016 hPa ( 20 °C)

Quality Level

Assay

≥99.0% (argenometric)

form

crystalline solid

potency

660 mg/kg LD50, oral (Rat)

pH

1 (5 g/L in H2O, acidic)

mp

148 °C

density

2.1 g/cm3 at 20 °C

storage temp.

2-30°C

InChI

1S/Cl5P/c1-6(2,3,4)5

InChI key

UHZYTMXLRWXGPK-UHFFFAOYSA-N

Application


  • Phosphorylation of Glycoluryl Derivatives: New organoelement phosphorus derivatives synthesized using glycoluril and phosphorus pentachloride under controlled conditions (Salkeeva et al., 2021).

  • Alcohol Amination and Cyclization: Catalyzed alcohol amination of diethanolamine by a Ru-PNP Pincer complex using phosphorus pentachloride to produce piperazine, demonstrating high conversion rates (van der Loo et al.).

  • Chemical Weapons Convention Relevance: Rapid GC-MS screening of chlorides relevant to the Chemical Weapons Convention, including phosphorus pentachloride, for monitoring prohibited chemical activities (Kuitunen et al., 2020).

  • Silicon Nanocrystal Functionalization: Phosphorus pentachloride used for the functionalization of silicon nanocrystals, enhancing their utility in advanced materials applications (Islam et al., 2017).


Analysis Note

Assay (argentometric): ≥ 99.0 %

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 2 Inhalation - Acute Tox. 4 Oral - Eye Dam. 1 - Skin Corr. 1B - STOT RE 2 Inhalation

Target Organs

Respiratory Tract

Supplementary Hazards

Storage Class Code

6.1B - Non-combustible, acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service