Skip to Content
Merck
All Photos(2)

Key Documents

8.22325

Sigma-Aldrich

4′-Hydroxyacetanilide

for synthesis

Synonym(s):

4′-Hydroxyacetanilide

Sign Into View Organizational & Contract Pricing

Select a Size

50 G
€40.80
250 G
€41.90
1 KG
€138.00
25 KG
€1,590.00

€40.80


Estimated to ship on30 May 2025


Request a Bulk Order

Select a Size

Change View
50 G
€40.80
250 G
€41.90
1 KG
€138.00
25 KG
€1,590.00

About This Item

Linear Formula:
CH3CONHC6H4(OH)
CAS Number:
Molecular Weight:
151.16
MDL number:
UNSPSC Code:
12352111
EC Index Number:
203-157-5
NACRES:
NA.22

€40.80


Estimated to ship on30 May 2025


Request a Bulk Order

Quality Level

Assay

≥99.0% (HPLC)

form

powder

autoignition temp.

540 °C

potency

1944 mg/kg LD50, oral (Rat)

expl. lim.

15 % (v/v)

pH

5.5-6.5 (20 °C in H2O, saturated solution)

mp

169-170 °C

solubility

14 g/L

density

1.293 g/cm3 at 21 °C

storage temp.

2-30°C

SMILES string

N(c1ccc(cc1)O)C(=O)C

InChI

1S/C8H9NO2/c1-6(10)9-7-2-4-8(11)5-3-7/h2-5,11H,1H3,(H,9,10)

InChI key

RZVAJINKPMORJF-UHFFFAOYSA-N

Application


  • A Review on the Properties and Uses of Paracetamol: This article provides a comprehensive overview of paracetamol, detailing its critical properties and varied uses in the pharmaceutical industry. (Ogemdi, 2019).

  • Development and Validation of a Chromatographic Method for Estimating Paracetamol: Discusses a method for simultaneous estimation of paracetamol and other compounds, highlighting its pharmacological significance. (Navneet Kumar et al., 2017).

  • Quality Assessment of Paracetamol Tablets: Evaluates the quality of different brands of paracetamol tablets in the Yemeni market, crucial for consumer safety. (Ali Alyahawi et al., 2018).

Analysis Note

Assay (HPLC, area%): ≥ 99.0 % (a/a)
Melting range (lower value): ≥ 168 °C
Melting range (upper value): ≤ 172 °C
Identity (IR): passes test

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

364.3 °F - Pensky-Martens closed cup

Flash Point(C)

184.6 °C - Pensky-Martens closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 3

1 of 3

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service