8.22101
Morpholinium chloride
for synthesis
Synonym(s):
Morpholinium chloride, Morpholine hydrochloride
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
Quality Level
potency
4580 mg/kg LD50, oral (Rat)
mp
174-178 °C
storage temp.
2-30°C
InChI
1S/C4H9NO.ClH/c1-3-6-4-2-5-1;/h5H,1-4H2;1H
InChI key
JXYZHMPRERWTPM-UHFFFAOYSA-N
Analysis Note
Assay (argentometric): ≥ 98.0 %
Melting range (lower value): ≥ 174 °C
Melting range (upper value): ≤ 178 °C
Sulfated ash: ≤ 0.20 %
Identity (IR): passes test
Melting range (lower value): ≥ 174 °C
Melting range (upper value): ≤ 178 °C
Sulfated ash: ≤ 0.20 %
Identity (IR): passes test
Storage Class Code
11 - Combustible Solids
WGK
WGK 2
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service