Skip to Content
Merck
All Photos(1)

Documents

8.21987

Sigma-Aldrich

Chloroethane

for synthesis

Synonym(s):

Chloroethane

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
64.51
MDL number:
UNSPSC Code:
12142100
EC Index Number:
200-830-5

vapor pressure

1342 hPa ( 20 °C)

form

gas

autoignition temp.

510 °C

expl. lim.

3.6-14.8 % (v/v)

bp

12 °C/1013 hPa

mp

-142 °C

transition temp

flash point -43 °C

solubility

5.8 g/L (slow decomposition)

density

0.92 g/cm3 at 6 °C

InChI

1S/C2H5Cl/c1-2-3/h2H2,1H3

InChI key

HRYZWHHZPQKTII-UHFFFAOYSA-N

General description

for synthesis

Application

Chloroethane for synthesis. CAS 75-00-3, molar mass 64.51 g/mol.

Analysis Note

Assay (GC, area%)≥ 99.0 % (a/a)

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Chronic 3 - Carc. 2 - Flam. Gas 1 - Press. Gas Liquefied gas

Storage Class Code

2A - Gases

WGK

WGK 2

Flash Point(C)

-50.0 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service