Skip to Content
Merck
All Photos(1)

Documents

8.21926

Sigma-Aldrich

Acetyl bromide

for synthesis

Synonym(s):

Acetyl bromide, Acetic acid bromide, Acetoxy bromide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C2H3BrO
CAS Number:
Molecular Weight:
122.95
MDL number:
UNSPSC Code:
12352106
EC Index Number:
208-061-7

vapor pressure

133 hPa ( 20 °C)

Quality Level

bp

76.7 °C/1013 mbar

mp

-96 °C

transition temp

flash point 177 °C

density

1.65 g/cm3 at 20 °C

InChI

1S/C2H3BrO/c1-2(3)4/h1H3

InChI key

FXXACINHVKSMDR-UHFFFAOYSA-N

Analysis Note

Assay (morpholine method): ≥ 97.0 %
Density (d 20 °C/ 4 °C): 1.650 - 1.670
Identity (IR): passes test

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service