Skip to Content
Merck
All Photos(1)

Key Documents

8.21222

Sigma-Aldrich

2,3-Dimethylaniline

for synthesis

Synonym(s):

2,3-Dimethylaniline, vic.-o-Xylidine

Sign Into View Organizational & Contract Pricing

Select a Size

5 ML
€25.90
250 ML
€41.20

€25.90


Estimated to ship on29 May 2025


Request a Bulk Order

Select a Size

Change View
5 ML
€25.90
250 ML
€41.20

About This Item

Linear Formula:
2,3-(CH3)2C6H3NH2
CAS Number:
Molecular Weight:
121.18
MDL number:
UNSPSC Code:
12352116
EC Index Number:
201-755-0
NACRES:
NA.22

€25.90


Estimated to ship on29 May 2025


Request a Bulk Order

vapor pressure

0.13 hPa ( 25 °C)

Quality Level

Assay

≥98.0% (GC)

form

liquid

autoignition temp.

545 °C

expl. lim.

1-2.7 % (v/v)

mp

2 °C

transition temp

flash point 96 °C

solubility

1.5 g/L

density

0.99 g/cm3 at 20 °C

storage temp.

2-30°C

SMILES string

Nc1c(c(ccc1)C)C

InChI

1S/C8H11N/c1-6-4-3-5-8(9)7(6)2/h3-5H,9H2,1-2H3

InChI key

VVAKEQGKZNKUSU-UHFFFAOYSA-N

General description

2,3-Dimethylaniline is an organic compound belonging to the class of aromatic amines. It is used as an organic building block in the synthesis of dyes and polymeric materials. Its amine group participates in polymerization reactions, including condensation and addition polymerization, with various monomers to form polymer chains. These polymers are crucial in producing plastics and resins, essential for advanced material manufacturing.

Analysis Note

Assay (GC, area%): ≥ 98.0 % (a/a)
Density (d 20 °C/ 4 °C): 0.991 - 0.993
Identity (IR): passes test
Due to its specific melting range the product may be solid, liquid, a solidified melt or a supercooled melt.

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Aquatic Chronic 2 - STOT RE 2

Storage Class Code

6.1A - Combustible, acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

204.8 °F - closed cup

Flash Point(C)

96 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service