Skip to Content
Merck
All Photos(1)

Key Documents

8.21197

Sigma-Aldrich

Triphenyl phosphate

for synthesis

Synonym(s):

Triphenyl phosphate, Phosphoric acid triphenyl ester

Sign Into View Organizational & Contract Pricing

Select a Size

100 G
€36.10
1 KG
€108.00

€36.10


Estimated to ship on28 May 2025


Request a Bulk Order

Select a Size

Change View
100 G
€36.10
1 KG
€108.00

About This Item

Linear Formula:
(C6H5O)3PO
CAS Number:
Molecular Weight:
326.28
MDL number:
UNSPSC Code:
12352108
EC Index Number:
204-112-2
NACRES:
NA.22

€36.10


Estimated to ship on28 May 2025


Request a Bulk Order

vapor pressure

<0.01 hPa ( 20 °C)

Quality Level

Assay

≥99.0% (GC)

form

solid

autoignition temp.

>500 °C

potency

3500 mg/kg LD50, oral (Rat)
>7900 mg/kg LD50, skin (Rabbit)

bp

220 °C/5 hPa

mp

48-50 °C

transition temp

flash point >230 °C

solubility

0.0019 g/L

bulk density

650 kg/m3

storage temp.

2-30°C

SMILES string

[P](=O)(Oc3ccccc3)(Oc2ccccc2)Oc1ccccc1

InChI

1S/C18H15O4P/c19-23(20-16-10-4-1-5-11-16,21-17-12-6-2-7-13-17)22-18-14-8-3-9-15-18/h1-15H

InChI key

XZZNDPSIHUTMOC-UHFFFAOYSA-N

Related Categories

Analysis Note

Assay (GC, area%): ≥ 99.0 % (a/a)
Melting range (lower value): ≥ 48 °C
Melting range (upper value): ≤ 50 °C
Identity (IR): passes test

Pictograms

Environment

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

428.0 °F - closed cup

Flash Point(C)

220 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service