Skip to Content
Merck
All Photos(1)

Key Documents

8.20833

Sigma-Aldrich

Dimethyl sulfide

for synthesis

Synonym(s):

Dimethyl sulfide, Methyl sulfide

Sign Into View Organizational & Contract Pricing

Select a Size

5 ML
€32.30
250 ML
€39.20
1 L
€84.40
45 KG
€1,500.00

€32.30


Estimated to ship on23 May 2025


Request a Bulk Order

Select a Size

Change View
5 ML
€32.30
250 ML
€39.20
1 L
€84.40
45 KG
€1,500.00

About This Item

Linear Formula:
(CH3)2S
CAS Number:
Molecular Weight:
62.13
MDL number:
UNSPSC Code:
12352113
EC Index Number:
200-846-2
NACRES:
NA.22

€32.30


Estimated to ship on23 May 2025


Request a Bulk Order

vapor pressure

530 hPa ( 25 °C)

Quality Level

Assay

≥99.0% (GC)

form

liquid

autoignition temp.

205 °C

potency

535 mg/kg LD50, oral (Rat)
>5000 mg/kg LD50, skin (Rabbit)

expl. lim.

2.2-19.7 % (v/v)

bp

37 °C/1013 hPa

mp

-98 °C

transition temp

flash point <-30 °C

solubility

7.28 g/L

density

0.85 g/cm3 at 20 °C

storage temp.

2-8°C

SMILES string

S(C)C

InChI

1S/C2H6S/c1-3-2/h1-2H3

InChI key

QMMFVYPAHWMCMS-UHFFFAOYSA-N

Application

  • Application of Synthetic Flavors in Zebrafish Rearing: The study details the effects of using synthetic flavors, including dimethyl sulfide, in the aquaculture of zebrafish, highlighting benefits in fish development and welfare. This application suggests potential for broader aquaculture practices (Conti et al., 2023).
  • Impact of Sulfur Fertilizers on Potato Tuber Flavor: Research explored the combined application of phosphorus and sulfur fertilizers on the flavor of potato tubers, noting that sulfur compounds like dimethyl sulfide can influence agricultural product quality (Li et al., 2023).
  • Two-Dimensional Platelets Assembly: A study demonstrated the use of small molecules including dimethyl sulfide for the living self-assembly of metastable and stable two-dimensional platelets, offering insights into novel materials engineering methods (Liao et al., 2023).
  • Enhancement of Volatile Flavor Compounds: Investigation on the effect of instant pressure drop treatment on the volatile flavor compounds in whole highland barley flour. The study emphasizes the role of dimethyl sulfide in enhancing food flavors through advanced processing techniques (Chen et al., 2023).
  • Role in Marine Biogeochemical Cycles: This research highlighted the contribution of fungi to the increased production of dimethylsulfoniopropionate (DMSP), a precursor to dimethyl sulfide, in kelp ecosystems, underscoring its importance in marine biogeochemical cycles (Perkins et al., 2023).

Analysis Note

Assay (GC, area%): ≥ 99.0 % (a/a)
Density (d 20 °C/ 4 °C): 0.847 - 0.849
Identity (IR): passes test

Pictograms

Flame

Signal Word

Danger

Hazard Statements

Hazard Classifications

Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

-32.8 °F - closed cup

Flash Point(C)

-36 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service